0 zł
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Poland
ContactUsLinkComponent
The adaptor is for use with Novex XCell I, II & SureLock® gel tanks when running ExpressPlus or SurePAGE gels (8x10cm) 1 * 2 items
GENSL00671
:
Genscript
The adaptor is for use with Novex XCell I, II & SureLock® gel tanks when running ExpressPlus or SurePAGE gels (8x10cm) 1 * 2 items
GENSL00671
:
Genscript
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Eppendorf® Protein LoBind® Microcentrifuge Tubes
: EPPENDORF
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
VWR®, Petri Dish, PS
: VWR Collection
VWR®, Cell Culture Flasks, Non Treated or Treated
: VWR Collection
VWR® PCR Tubes, 0,2 ml
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Sampling Spoons
: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Hygromycin B 100 mg/ml in water (from Streptomyces hygroscopicus) , 90% (by HPLC), sterile filtered
: Enzo Life Sciences