rac-1,2-bis-Palmitoyl-3-chloropropanediol 1 * 10 mg
CAYM9003677-10
Frequently Bought Together
Methanol, anhydrous ≥99.9%
: Thermo Fisher Scientific
Isoamyl alcohol 98%, pure
: Thermo Fisher Scientific
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
: PanReac AppliChem
Acetonitrile, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
: Honeywell Chemicals
Petroleum spirit, 30…40 °C, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
n-Decane ≥99%, Reagent Grade
: Honeywell Chemicals
N,N-Dimethylformamide, anhydrous ≥99.8%
: Thermo Fisher Scientific
Methanol for DNA and peptides synthesis
: POCH AVANTOR BRAND
ETYLU OCTAN CZDA, ACS OP. 18 DM3 1 * 18 L
: POCH AVANTOR BRAND
Cyclohexanone, Sigma-Aldrich®
: Merck
4-(Methylamino)phenol sulphate 99%, extra pure
: Thermo Fisher Scientific