UNC0064-12 1 * 10 mg
CAYM33555-10
Frequently Bought Together
Benzyl alcohol ≥99.0% analytical reagent
: VWR Chemicals
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
: PanReac AppliChem
Acetonitrile ≥99.7%, Environmental Grade
: Thermo Fisher Scientific
Dichloromethane 99.8% stabilised for HPLC
: Thermo Fisher Scientific
Trifluoroacetic Acid for UV 1 * 100 mL
: PanReac AppliChem
Tetrahydrofuran 99.8% unstabilised for HPLC
: Thermo Fisher Scientific
n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
: Honeywell Chemicals
N,N-Dimethylformamide ≥99.8% ACS for spectroscopy
: Thermo Fisher Scientific
Acetone (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
: PanReac AppliChem
N,N-Dimethylformamide ≥99.8% ACS
: Thermo Fisher Scientific
Acetone 99.6% ACS
: Thermo Fisher Scientific
Acetic acid ≥99%, pure
: POCH AVANTOR BRAND