Order Entry
Poland
ContactUsLinkComponent
SARS-CoV-2 Nucleocapsid protein His Tag (B.1.1.529/Omicron) (NUN-C52Ht) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 419 (Accession # QHO62115.1 (P13L ERS31-33del R203K G204R)). The mutations are identified on the SARS-Co ar 1 * 1 mg
Catalog # ACRBNUN-C52HT-1MG
Supplier:  ACRO Biosystems
SARS-CoV-2 Nucleocapsid protein His Tag (B.1.1.529/Omicron) (NUN-C52Ht) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 419 (Accession # QHO62115.1 (P13L ERS31-33del R203K G204R)). The mutations are identified on the SARS-Co ar 1 * 1 mg
Catalog # ACRBNUN-C52HT-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together