Order Entry
Poland
Orders LinkContactUsLinkComponent
VWR®, Septa ND11 | Avantor

VWR®, Septa ND11

Red rubber/beige PTFE, 45° shore A, 1,0 mm
numer katalogowy 548-3302A
Dostawca:   VWR Collection
VWR®, Septa ND11
numer katalogowy 548-3302A
Dostawca:  VWR Collection
VWR®, Septa ND11
VWR®, Septa ND11

Red rubber/beige PTFE, 45° shore A, 1,0 mm

Przejdź do:
Price
914 zł
Zaloguj się dla dostępności
1 * 1.000 SZT
Product Details & Documents

Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.

  • Fits for a broad range of caps
  • Easy exchange
  • Different hardness and septa thickness available

Septa choice depends on the application.

Dane techniczne
  • Hardness:
    45° shore A
  • Septum:
    Red rubber/beige PTFE
  • Seq:
    10
  • Thickness:
    1,0 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.