Order Entry
Poland
ContactUsLinkComponent
Nipah virus Fusion glycoprotein, His Tag (FUN-N52H3) is expressed from human 293 cells (HEK293). It contains AA Ile 27 - Ser 487 (Ac cession # Q9IH63-1). 1 * 50 µG
Catalog # ACRBFUN-N52H3-50UG
Supplier:  ACROBIOSYSTEMS
Nipah virus Fusion glycoprotein, His Tag (FUN-N52H3) is expressed from human 293 cells (HEK293). It contains AA Ile 27 - Ser 487 (Ac cession # Q9IH63-1). 1 * 50 µG
Catalog # ACRBFUN-N52H3-50UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together