Order Entry
Poland
ContactUsLinkComponent
Human CD3E&CD3D Heterodimer Protein (CDD-H52Wa) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (CD3E) & Phe 22 - Ala 105 (CD3D) (Accession # P07766-1 (CD3E) & P04234-1 (CD3D)). 1 * 100 µG
Catalog # ACRBCDD-H52WA100UG
Supplier:  ACRO Biosystems
Human CD3E&CD3D Heterodimer Protein (CDD-H52Wa) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (CD3E) & Phe 22 - Ala 105 (CD3D) (Accession # P07766-1 (CD3E) & P04234-1 (CD3D)). 1 * 100 µG
Catalog # ACRBCDD-H52WA100UG
Supplier:  ACRO Biosystems
Frequently Bought Together