Human CD3E&CD3D Heterodimer Protein, Fc Tag&Fc Tag (CDD-H5255) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - A sp 126 (CD3E) & Phe 22 - Ala 105 (CD3D) (Accession # P07766-1 (CD3E) & P04234-1 (CD3D)). 1 * 1 mg
Catalog # ACRBCDD-H5255-1MG
Dostawca: ACRO Biosystems
Frequently Bought Together