Order Entry
Poland
ContactUsLinkComponent
Biotinylated SARS-CoV-2 Spike NTD, His,Avitag (SPD-C82H3) is expressed from human 293 cells (HEK293). It contains AA Ser 13 - Leu 30 3 (Accession # QHD43416.1). 1 * 200 µG
Catalog # ACRBSPD-C82H3200UG
Supplier:  ACRO Biosystems
Biotinylated SARS-CoV-2 Spike NTD, His,Avitag (SPD-C82H3) is expressed from human 293 cells (HEK293). It contains AA Ser 13 - Leu 30 3 (Accession # QHD43416.1). 1 * 200 µG
Catalog # ACRBSPD-C82H3200UG
Supplier:  ACRO Biosystems
Frequently Bought Together