Rat IL-2RB&IL-2RA&IL-2RG, His Tag&Twin-Strep Tag (ILG-R5223) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Glu 239 (IL-2RB) & Glu 22 - Gln 235 (IL-2RA) & Ser 25 - Ala 262 (IL-2RG) (Accession # P26896-1 (IL-2RB) & P26897-1 (IL- 1 * 100 µG
Catalog # ACRBILG-R5223100UG
Supplier: ACRO Biosystems
Frequently Bought Together