Order Entry
Poland
ContactUsLinkComponent
Cynomolgus Tissue Factor, His Tag (TF3-C52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 33 - Glu 252 (Accession # A0A2K5VX02-1). 1 * 500 µG
Catalog # ACRBTF3-C52H3500UG
Supplier:  ACRO Biosystems
Cynomolgus Tissue Factor, His Tag (TF3-C52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 33 - Glu 252 (Accession # A0A2K5VX02-1). 1 * 500 µG
Catalog # ACRBTF3-C52H3500UG
Supplier:  ACRO Biosystems
Frequently Bought Together