Order Entry
Poland
ContactUsLinkComponent
Mouse PVRIG, Mouse IgG2a Fc Tag (PVG-M5253) is expressed from human 293 cells (HEK293). It contains AA Ser 35 - Asp 165 (Accession # A0A1B0GS01-1). 1 * 100 µG
Catalog # ACRBPVG-M5253100UG
Supplier:  ACRO Biosystems
Mouse PVRIG, Mouse IgG2a Fc Tag (PVG-M5253) is expressed from human 293 cells (HEK293). It contains AA Ser 35 - Asp 165 (Accession # A0A1B0GS01-1). 1 * 100 µG
Catalog # ACRBPVG-M5253100UG
Supplier:  ACRO Biosystems
Frequently Bought Together