Order Entry
Poland
ContactUsLinkComponent
Canine TROP-2, His Tag (TR2-C52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 39 - Thr 281 (Accession # XP_85284 2.5). 1 * 100 µG
Catalog # ACRBTR2-C52H4100UG
Supplier:  ACRO Biosystems
Canine TROP-2, His Tag (TR2-C52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 39 - Thr 281 (Accession # XP_85284 2.5). 1 * 100 µG
Catalog # ACRBTR2-C52H4100UG
Supplier:  ACRO Biosystems
Frequently Bought Together