Order Entry
Poland
ContactUsLinkComponent
PE-Labeled Human CD7, His Tag (CD7-HP2E3) is produced via site-specific conjugation of PE to Human CD7 (26-180), His Tag under opti mal conditions with a proprietary technology. PE-Labeled Human CD7, His Tag (CD7-HP2E3) is expressed from human 293 ce 1 * 25 Tests
Catalog # ACRBCD7HP2E325TST
Supplier:  ACRO Biosystems
PE-Labeled Human CD7, His Tag (CD7-HP2E3) is produced via site-specific conjugation of PE to Human CD7 (26-180), His Tag under opti mal conditions with a proprietary technology. PE-Labeled Human CD7, His Tag (CD7-HP2E3) is expressed from human 293 ce 1 * 25 Tests
Catalog # ACRBCD7HP2E325TST
Supplier:  ACRO Biosystems
Frequently Bought Together