Order Entry
Poland
ContactUsLinkComponent
Cynomolgus CLEC4C, His Tag (CLC-C52H4) is expressed from human 293 cells (HEK293). It contains AA Tyr 48 - Ile 212 (Accession # A0A2 K5UWP4-1). 1 * 1 mg
Catalog # ACRBCLC-C52H4-1MG
Supplier:  ACROBIOSYSTEMS
Cynomolgus CLEC4C, His Tag (CLC-C52H4) is expressed from human 293 cells (HEK293). It contains AA Tyr 48 - Ile 212 (Accession # A0A2 K5UWP4-1). 1 * 1 mg
Catalog # ACRBCLC-C52H4-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together