Order Entry
Poland
Orders LinkContactUsLinkComponent
Pterostilbene

Pterostilbene

Bottle, 25 g
Catalog # APOSBIP1700-25G
Corrosive
Environment
Supplier:   Apollo Scientific
Pterostilbene
Catalog # APOSBIP1700-25G
Supplier:  Apollo Scientific
CAS Number:  537-42-8
Pterostilbene

Bottle, 25 g

CAS Number:  537-42-8
Price
707 zł
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  (E)-4-(3,5-Dimethoxystyryl)phenol, 4-[(E)-2-(3,5-Dimethoxyphenyl)ethenyl]phenol
  • CAS Number:537-42-8
  • Formula:C₁₆H₁₆O₃
  • MDL Number:MFCD00238710
  • Molecular Weight:256.2964 g/mol
  • Storage Temperature:Refrigerator
  • UN:3077
  • ADR:9+EHS,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.