Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PEth 16:0/18:2 solution, 1.0 mg/mL in methanol (as free phosphate), certified reference material, ampule of 1 mL, Cerilliant® 1 * 1 mL
Catalog # P-115-1ML
Supplier:
Merck
Expand image
PEth 16:0/18:2 solution, 1.0 mg/mL in methanol (as free phosphate), certified reference material, ampule of 1 mL, Cerilliant® 1 * 1 mL
Catalog # P-115-1ML
Supplier:
Merck
Frequently Bought Together
Gold standard solution, 1,000 mg/L Au in 2% hydrochloric acid standard for ICP
Supplier: CPACHEM
Drug and metabolite standards, 7-Hydroxyquetiapine solution, (CRM), Supelco®
Supplier: Merck
VISCOSITY STANDARD, GENERAL PURPOSE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 55.36 MPA.S (25 °C) 1 * 500 mL
Supplier: PARAGON SCIENTIFIC
Chitosan, Reference Standard
Supplier: USP
Magnesium Sulfate, Reference Standard
Supplier: USP
Supelco® Organic Reference Standard, Palmitoleic acid
Supplier: Merck
Valeric acid, Supelco®
Supplier: Merck
Calcium standard solution, 10000 mg/L Ca in 5% nitric acid standard for ICP
Supplier: CPACHEM
LOFEXIDINE HCL 1 * 1 mL
Supplier: Merck
Copper Standard Solution, 1000 mg/L Cu 1 * 100 mL
Supplier: Merck
Organic reference standard, Trimethylamine hydrochloride, analytical standard, Supelco®
Supplier: Merck