Selective, non-competitive and reversible IMPDH inhibitor. Immunosuppressant agent. 1 * 250 mg
ABCAAB143013-250
: Abcam
Selective, non-competitive and reversible IMPDH inhibitor. Immunosuppressant agent. 1 * 250 mg
ABCAAB143013-250
: Abcam
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection