Order Entry
Poland
Orders LinkContactUsLinkComponent
Methyl-4-hydroxybenzoate 98%

Methyl-4-hydroxybenzoate 98%

Bottle, 100 g
Catalog # APOSOR922886-100G
Supplier:   Apollo Scientific
Methyl-4-hydroxybenzoate 98%
Catalog # APOSOR922886-100G
Supplier:  Apollo Scientific
CAS Number:  99-76-3
Methyl-4-hydroxybenzoate 98%

Bottle, 100 g

CAS Number:  99-76-3
Price
107 zł
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate
  • CAS Number:99-76-3
  • Formula:HOC₆H₄CO₂CH₃
  • MDL Number:MFCD00002352
  • Molecular Weight:152.15 g/mol
  • Melting Pt:125…128 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications