3-(4-Chlorophenoxy)butan-2-one, tech
Bottle, 1 g
numer katalogowy APOSOR33209-1G
Dostawca: Apollo Scientific
3-(4-Chlorophenoxy)butan-2-one, tech
numer katalogowy APOSOR33209-1G
Dostawca: Apollo Scientific
Numer CAS: 3782-11-4
Przejdź do:
Product Details & Documents
- Numer CAS:3782-11-4
- Wzór:C₁₀H₁₁ClO₂
- Numer MDL:MFCD00243668
- Molecular Weight:198.646 g/mol
- Temperatura przechowywania:Ambient
Dane techniczne
Dane techniczne
- Pack type:Bottle
- Pk:1 g
Dane techniczne
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



