Order Entry
Poland
Orders LinkContactUsLinkComponent
 

1,8-Octanediol 98%

Bottle, 1 kg
Catalog # APOSOR2431-1KG
Irritant
Supplier:   Apollo Scientific
1,8-Octanediol 98%
Catalog # APOSOR2431-1KG
Supplier:  Apollo Scientific
CAS Number:  629-41-4
1,8-Octanediol 98%

Bottle, 1 kg

CAS Number:  629-41-4
Price
1 700 zł
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  1,8-Dihydroxy octane
  • CAS Number:629-41-4
  • Formula:C₈H₁₈O₂
  • MDL Number:MFCD00002989
  • Molecular Weight:146.23 g/mol
  • Boiling Pt:71…73 °C/20mmHg
  • Melting Pt:59…61 °C
  • Flash Pt:148 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg

Specifications