Order Entry
Poland
Orders LinkContactUsLinkComponent
3,3'-Dihydroxybenzidine 97%

3,3'-Dihydroxybenzidine 97%

Bottle, 1 g
Catalog # APOSOR912142-1G
Irritant
Health
Supplier:   Apollo Scientific
3,3'-Dihydroxybenzidine 97%
Catalog # APOSOR912142-1G
Supplier:  Apollo Scientific
CAS Number:  2373-98-0
3,3'-Dihydroxybenzidine 97%

Bottle, 1 g

CAS Number:  2373-98-0
Price
107 zł
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  4,4'-Diamino-3,3'-dihydroxybiphenyl, 3,3'-Dihydroxy-4,4'-diaminobiphenyl, 4,4'-Diamino-[1,1'-biphenyl]-3,3'-diol, Benzidine-3,3'-diol
  • CAS Number:2373-98-0
  • Formula:C₁₂H₁₂N₂O₂
  • MDL Number:MFCD00039149
  • Molecular Weight:216.24 g/mol
  • Melting Pt:292 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.