Złóż zamówienie
Poland
Orders LinkContactUsLinkComponent
 

3-(3-Fluorophenoxy)butan-2-one, tech

Bottle, 1 g
Catalog # APOSPC27983-1G
Supplier:   Apollo Scientific
3-(3-Fluorophenoxy)butan-2-one, tech
Catalog # APOSPC27983-1G
Supplier:  Apollo Scientific
CAS Number:  306979-64-6
3-(3-Fluorophenoxy)butan-2-one, tech

Bottle, 1 g

CAS Number:  306979-64-6
Price
7 630 zł
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:306979-64-6
  • Formula:C₁₀H₁₁FO₂
  • MDL Number:MFCD00243584
  • Molecular Weight:182.192 g/mol
  • Storage Temperature:Temperatura otoczenia
Specifications

Specifications

  • Rodzaj opakowania:
    Bottle
  • Ilość:
    1 g

Specifications