Order Entry
Poland
ContactUsLinkComponent
Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1 is produced from a hybridoma resulting from fusion of SP20 myeloma and B-lymphocytes obtained from a mouse immunized with FMC63 scFv. 1 * 1.000 Tests
Catalog # ACRBFM3Y451000TEST
Supplier:  ACRO Biosystems
Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1 is produced from a hybridoma resulting from fusion of SP20 myeloma and B-lymphocytes obtained from a mouse immunized with FMC63 scFv. 1 * 1.000 Tests
Catalog # ACRBFM3Y451000TEST
Supplier:  ACRO Biosystems
Frequently Bought Together