Order Entry
Poland
ContactUsLinkComponent
Biotinylated Human CD39, His,Avitag MALS verified CD9-H82E5 is expressed from human 293 cells HEK293. It contains AA Thr 38 - Val 47 8 Accession P49961-1. 1 * 200 µG
Catalog # ACRBCD9-H82E5200UG
Supplier:  ACRO Biosystems
Biotinylated Human CD39, His,Avitag MALS verified CD9-H82E5 is expressed from human 293 cells HEK293. It contains AA Thr 38 - Val 47 8 Accession P49961-1. 1 * 200 µG
Catalog # ACRBCD9-H82E5200UG
Supplier:  ACRO Biosystems
Frequently Bought Together