Order Entry
Poland
ContactUsLinkComponent
Human FCGRTB2M Heterodimer Protein, His Tag SPR  BLI verified FCN-H52W7 is expressed from human 293 cells HEK293. It contains AA Ala 24 - Ser 297 FCGRT Ile 21 - Met 119 B2M Accession P55899-1 FCGRT P61769-1 B2M. 1 * 50 µG
  ACRBFCN-H52W7-50UG
Human FCGRTB2M Heterodimer Protein, His Tag SPR  BLI verified FCN-H52W7 is expressed from human 293 cells HEK293. It contains AA Ala 24 - Ser 297 FCGRT Ile 21 - Met 119 B2M Accession P55899-1 FCGRT P61769-1 B2M. 1 * 50 µG
  ACRBFCN-H52W7-50UG
Frequently Bought Together