Biotinylated Human CD27 Ligand, His,Avitag,Flag Tag active trimer MALS verified CDL-H82D7 is expressed from human 293 cells HEK293. It contains AA Ser 52 - Pro 193 Accession P32970-1 trimer Design. 1 * 25 µG
Catalog # ACRBCDL-H82D7-25UG
Supplier: ACRO Biosystems
Frequently Bought Together