Order Entry
Poland
ContactUsLinkComponent
Human IL-15, His Tag IL5-H52H8 is expressed from human 293 cells HEK293. It contains AA Asn 49 - Ser 162 Accession P40933-1. 1 * 50 µG
Catalog # ACRBIL5-H52H8-50UG
Supplier:  ACRO Biosystems
Human IL-15, His Tag IL5-H52H8 is expressed from human 293 cells HEK293. It contains AA Asn 49 - Ser 162 Accession P40933-1. 1 * 50 µG
Catalog # ACRBIL5-H52H8-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together