Order Entry
Poland
ContactUsLinkComponent
Rat ACE2, His Tag AC2-R5246 is expressed from human 293 cells HEK293. It contains AA Gln 18 - Thr 740 Accession Q5EGZ1-1. 1 * 1 mg
Catalog # ACRBAC2-R5246-1MG
Supplier:  ACRO Biosystems
Rat ACE2, His Tag AC2-R5246 is expressed from human 293 cells HEK293. It contains AA Gln 18 - Thr 740 Accession Q5EGZ1-1. 1 * 1 mg
Catalog # ACRBAC2-R5246-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together