Order Entry
Poland
ContactUsLinkComponent
SARS-CoV-2 S1 protein, Mouse IgG2a Fc Tag S1N-C5257 is expressed from human 293 cells HEK293. It contains AA Val 16 - Arg 685 Access ion QHD43416.1. 1 * 100 µG
Catalog # ACRBS1N-C5257100UG
Supplier:  ACRO Biosystems
SARS-CoV-2 S1 protein, Mouse IgG2a Fc Tag S1N-C5257 is expressed from human 293 cells HEK293. It contains AA Val 16 - Arg 685 Access ion QHD43416.1. 1 * 100 µG
Catalog # ACRBS1N-C5257100UG
Supplier:  ACRO Biosystems
Frequently Bought Together