Order Entry
Poland
ContactUsLinkComponent
SARS-CoV-2 S2 protein, His Tag S2N-C52H5 is expressed from human 293 cells HEK293. It contains AA Ser 686 - Pro 1213 Accession QHD4 3416.1. Proline substitutions F817P, A892P, A899P, A942P, K986P, V987P are introduced to prevent the formation of aggr 1 * 100 µG
Catalog # ACRBS2N-C52H5100UG
Supplier:  ACRO Biosystems
SARS-CoV-2 S2 protein, His Tag S2N-C52H5 is expressed from human 293 cells HEK293. It contains AA Ser 686 - Pro 1213 Accession QHD4 3416.1. Proline substitutions F817P, A892P, A899P, A942P, K986P, V987P are introduced to prevent the formation of aggr 1 * 100 µG
Catalog # ACRBS2N-C52H5100UG
Supplier:  ACRO Biosystems
Frequently Bought Together