Order Entry
Poland
ContactUsLinkComponent
SARS-CoV-2 Papain-like Protease, His Tag PAE-C5148 is expressed from E.coli cells. It contains AA Glu 746 - Lys 1060 Accession YP_0 09725299.1. 1 * 100 µG
Catalog # ACRBPAE-C5148100UG
Supplier:  ACRO Biosystems
SARS-CoV-2 Papain-like Protease, His Tag PAE-C5148 is expressed from E.coli cells. It contains AA Glu 746 - Lys 1060 Accession YP_0 09725299.1. 1 * 100 µG
Catalog # ACRBPAE-C5148100UG
Supplier:  ACRO Biosystems
Frequently Bought Together