Order Entry
Poland
ContactUsLinkComponent
SARS-CoV-2 NSP8, His Tag NS8-C5149 is expressed from E.coli cells. It contains AA Ala 1 - Gln 198 Accession YP_009725304.1. 1 * 1 mg
Catalog # ACRBNS8-C5149-1MG
Supplier:  ACRO Biosystems
SARS-CoV-2 NSP8, His Tag NS8-C5149 is expressed from E.coli cells. It contains AA Ala 1 - Gln 198 Accession YP_009725304.1. 1 * 1 mg
Catalog # ACRBNS8-C5149-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together