Order Entry
Poland
ContactUsLinkComponent
SARS-CoV-2 NSP1, His Tag NS1-C51H7 is expressed from E.coli cells. It contains AA Met 1 - Gly 180 Accession YP_009725297.1. 1 * 100 µG
Catalog # ACRBNS1-C51H7100UG
Supplier:  ACRO Biosystems
SARS-CoV-2 NSP1, His Tag NS1-C51H7 is expressed from E.coli cells. It contains AA Met 1 - Gly 180 Accession YP_009725297.1. 1 * 100 µG
Catalog # ACRBNS1-C51H7100UG
Supplier:  ACRO Biosystems
Frequently Bought Together