Order Entry
Poland
ContactUsLinkComponent
SARS-CoV-2 NSP16NSP10 Heterodimer Protein, His TagTwin Strep Tag NS0-C51W3 is expressed from E.coli cells. It contains AA Ser 1 - As n 298 NSP16 Ala 1 - Gln 139 NSP10 Accession YP_009725311.1 NSP16 YP_009725306.1 NSP10. 1 * 1 mg
Catalog # ACRBNS0-C51W3-1MG
Supplier:  ACRO Biosystems
SARS-CoV-2 NSP16NSP10 Heterodimer Protein, His TagTwin Strep Tag NS0-C51W3 is expressed from E.coli cells. It contains AA Ser 1 - As n 298 NSP16 Ala 1 - Gln 139 NSP10 Accession YP_009725311.1 NSP16 YP_009725306.1 NSP10. 1 * 1 mg
Catalog # ACRBNS0-C51W3-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together