Recombinant Streptavidin STN-N5116 is expressed from E.coli cells. 1 * 5 mg
Catalog # ACRBSTN-N5116-5MG
Supplier: ACRO Biosystems
Recombinant Streptavidin STN-N5116 is expressed from E.coli cells. 1 * 5 mg
Catalog # ACRBSTN-N5116-5MG
Supplier: ACRO Biosystems
Frequently Bought Together
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR®, Microscope Slides
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, LTS® Compatible Pipette Tips
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
Granular dessicant, Sicapent® with indicator, Supelco®
Supplier: Merck
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Bovine Serum Albumin (BSA), Fraction V, MP Biomedicals
Supplier: MP Biomedicals