Order Entry
Poland
ContactUsLinkComponent
Mouse FGL1, Fc Tag FG1-M5258 is expressed from human 293 cells HEK293. It contains AA Leu 23 - Ile 314 Accession Q71KU9-1. 1 * 1 mg
Catalog # ACRBFG1-M5258-1MG
Supplier:  ACRO Biosystems
Mouse FGL1, Fc Tag FG1-M5258 is expressed from human 293 cells HEK293. It contains AA Leu 23 - Ile 314 Accession Q71KU9-1. 1 * 1 mg
Catalog # ACRBFG1-M5258-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together