Order Entry
Poland
ContactUsLinkComponent
SARS-CoV-2 Nucleocapsid protein, His Tag NUN-C51H2 is expressed from E.coli cells. It contains AA Met 1 - Ala 419 Accession QHO6211 5.1. 1 * 100 mg
Catalog # ACRBNUN-C51H2100MG
Supplier:  ACRO Biosystems
SARS-CoV-2 Nucleocapsid protein, His Tag NUN-C51H2 is expressed from E.coli cells. It contains AA Met 1 - Ala 419 Accession QHO6211 5.1. 1 * 100 mg
Catalog # ACRBNUN-C51H2100MG
Supplier:  ACRO Biosystems
Frequently Bought Together