Order Entry
Poland
ContactUsLinkComponent
The Streptavidin Coated Plates, Clear, 96-Well is pre-coated with Streptavidin tetramer protein and blocked with BSA, it is ready-to -use polystyrene plate, which can be used for binding biotinylated proteins and antibodies, or probes for ELISA and o 1 * 1 items
Catalog # ACRBSP-11-1PLATE
Supplier:  ACROBIOSYSTEMS
The Streptavidin Coated Plates, Clear, 96-Well is pre-coated with Streptavidin tetramer protein and blocked with BSA, it is ready-to -use polystyrene plate, which can be used for binding biotinylated proteins and antibodies, or probes for ELISA and o 1 * 1 items
Catalog # ACRBSP-11-1PLATE
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together