Order Entry
Poland
ContactUsLinkComponent
The biotinylated Anti-SARS-CoV-2 Spike RBD Antibody was conjugated to streptavidin magnetic beads. This pre-coupled magnetic bead pr oduct can capture the SARS-CoV-2 S1 proteinSARS-CoV-2 Spike RBD from various assay systems. The beads are in uniform 1 * 10 mg
Catalog # ACRBMBS-K014-10MG
Supplier:  ACRO Biosystems
The biotinylated Anti-SARS-CoV-2 Spike RBD Antibody was conjugated to streptavidin magnetic beads. This pre-coupled magnetic bead pr oduct can capture the SARS-CoV-2 S1 proteinSARS-CoV-2 Spike RBD from various assay systems. The beads are in uniform 1 * 10 mg
Catalog # ACRBMBS-K014-10MG
Supplier:  ACRO Biosystems
Frequently Bought Together