Order Entry
Poland
ContactUsLinkComponent
Biotinylated Human CD229, His,Avitag CD9-H82E6 is expressed from human 293 cells HEK293. It contains AA Lys 48 - Lys 454 Accession Q9HBG7-1. 1 * 200 µG
Catalog # ACRBCD9-H82E6200UG
Supplier:  ACRO Biosystems
Biotinylated Human CD229, His,Avitag CD9-H82E6 is expressed from human 293 cells HEK293. It contains AA Lys 48 - Lys 454 Accession Q9HBG7-1. 1 * 200 µG
Catalog # ACRBCD9-H82E6200UG
Supplier:  ACRO Biosystems
Frequently Bought Together