Order Entry
Poland
ContactUsLinkComponent
Biotinylated S1 protein NTD, His,Avitag™ S1D-C52E2 is expressed from human 293 cells HEK293. It contains AA Val 16 - Phe 318 Accessi on QHD43416.1. 1 * 25 µG
Catalog # ACRBS1D-C52E2-25UG
Supplier:  ACRO Biosystems
Biotinylated S1 protein NTD, His,Avitag™ S1D-C52E2 is expressed from human 293 cells HEK293. It contains AA Val 16 - Phe 318 Accessi on QHD43416.1. 1 * 25 µG
Catalog # ACRBS1D-C52E2-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together