PE-Labeled Human SLAMF7, His Tag SL7-HP2H3 is produced via conjugation of PE to Human SLAMF7, His Tag under optimal conditions. Huma n SLAMF7, His Tag is expressed from human 293 cells HEK293. It contains AA Ser 23 - Met 226 Accession Q9NQ25-1. 1 * 200 Tests
Catalog # ACRBSL7HP2H3200TST
Supplier: ACROBIOSYSTEMS
Frequently Bought Together