Order Entry
Poland
ContactUsLinkComponent
Biotinylated Cynomolgus CD3ECD3G Heterodimer Protein, His,AvitagFlag Tag CDG-C8219 is expressed from human 293 cells HEK293. It cont ains AA Gln 22 - Asp 117 CD3E Gln 23 - Thr 113 CD3G Accession Q95LI5-1 CD3E Q95LI7-1 CD3G. 1 * 200 µG
Catalog # ACRBCDG-C8219200UG
Supplier:  ACRO Biosystems
Biotinylated Cynomolgus CD3ECD3G Heterodimer Protein, His,AvitagFlag Tag CDG-C8219 is expressed from human 293 cells HEK293. It cont ains AA Gln 22 - Asp 117 CD3E Gln 23 - Thr 113 CD3G Accession Q95LI5-1 CD3E Q95LI7-1 CD3G. 1 * 200 µG
Catalog # ACRBCDG-C8219200UG
Supplier:  ACRO Biosystems
Frequently Bought Together