Order Entry
Poland
ContactUsLinkComponent
SARS-CoV-2 S protein, His Tag SPN-C52Hd is expressed from human 293 cells HEK293. It contains AA Val 16 - Pro 1213 F817P, A892P, A89 9P, A942P,K986P,V987P Accession QHD43416.1 F817P, A892P, A899P, A942P,K986P,V987P. Proline substitutions F817P, A89 P 1 * 500 µG
  ACRBSPN-C52HD500UG
SARS-CoV-2 S protein, His Tag SPN-C52Hd is expressed from human 293 cells HEK293. It contains AA Val 16 - Pro 1213 F817P, A892P, A89 9P, A942P,K986P,V987P Accession QHD43416.1 F817P, A892P, A899P, A942P,K986P,V987P. Proline substitutions F817P, A89 P 1 * 500 µG
  ACRBSPN-C52HD500UG
Frequently Bought Together