Order Entry
Poland
ContactUsLinkComponent
Rat CD3ECD3D Heterodimer Protein, His TagFlag Tag CDD-R52D8 is expressed from human 293 cells HEK293. It contains AA Gln 21 - Asp 10 3 CD3E Phe 22 - Gly 106 CD3D Accession D4A5M2-1 CD3E P19377-1 CD3D. 1 * 1 mg
Catalog # ACRBCDD-R52D8-1MG
Supplier:  ACRO Biosystems
Rat CD3ECD3D Heterodimer Protein, His TagFlag Tag CDD-R52D8 is expressed from human 293 cells HEK293. It contains AA Gln 21 - Asp 10 3 CD3E Phe 22 - Gly 106 CD3D Accession D4A5M2-1 CD3E P19377-1 CD3D. 1 * 1 mg
Catalog # ACRBCDD-R52D8-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together