Order Entry
Poland
ContactUsLinkComponent
SARS-CoV-2 S1 protein NTD HV69-70del, Y144del, His Tag S1D-C52Hd is expressed from human 293 cells HEK293. It contains AA Ser 13 - L eu 303 Accession QHD43416.1 HV69-70del, Y144del. 1 * 1 mg
Catalog # ACRBS1D-C52HD-1MG
Supplier:  ACRO Biosystems
SARS-CoV-2 S1 protein NTD HV69-70del, Y144del, His Tag S1D-C52Hd is expressed from human 293 cells HEK293. It contains AA Ser 13 - L eu 303 Accession QHD43416.1 HV69-70del, Y144del. 1 * 1 mg
Catalog # ACRBS1D-C52HD-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together