Order Entry
Poland
ContactUsLinkComponent
SARS-CoV-2 S1 protein, His Tag S1N-C52Hr is expressed from human 293 cells HEK293. It contains AA Val 16 - Arg 685 Accession QHD434 16.1 HV69-70del, Y144del, N501Y, A570D, D614G, P681H. 1 * 1 mg
Catalog # ACRBS1N-C52HR-1MG
Supplier:  ACRO Biosystems
SARS-CoV-2 S1 protein, His Tag S1N-C52Hr is expressed from human 293 cells HEK293. It contains AA Val 16 - Arg 685 Accession QHD434 16.1 HV69-70del, Y144del, N501Y, A570D, D614G, P681H. 1 * 1 mg
Catalog # ACRBS1N-C52HR-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together