Order Entry
Poland
ContactUsLinkComponent
SARS-CoV-2 S1 protein NTD, His Tag S1D-C52He is expressed from human 293 cells HEK293. It contains AA Ser 13 - Leu 303 Accession QH D43416.1 L18F, T20N, P26S, D138Y, R190S. 1 * 100 µG
Catalog # ACRBS1D-C52HE100UG
Supplier:  ACRO Biosystems
SARS-CoV-2 S1 protein NTD, His Tag S1D-C52He is expressed from human 293 cells HEK293. It contains AA Ser 13 - Leu 303 Accession QH D43416.1 L18F, T20N, P26S, D138Y, R190S. 1 * 100 µG
Catalog # ACRBS1D-C52HE100UG
Supplier:  ACRO Biosystems
Frequently Bought Together