Order Entry
Poland
ContactUsLinkComponent
SARS-CoV-2 S2 protein T716I, S982A, D1118H, His Tag S2N-C52Hd is expressed from human 293 cells HEK293. It contains AA Ser 686 - Pro 1213 Accession QHD43416.1 T716I, S982A, D1118H, F817P, A892P, A899P, A942P, K986P, V987P. The recombinant protein i m 1 * 1 mg
Catalog # ACRBS2N-C52HD-1MG
Supplier:  ACRO Biosystems
SARS-CoV-2 S2 protein T716I, S982A, D1118H, His Tag S2N-C52Hd is expressed from human 293 cells HEK293. It contains AA Ser 686 - Pro 1213 Accession QHD43416.1 T716I, S982A, D1118H, F817P, A892P, A899P, A942P, K986P, V987P. The recombinant protein i m 1 * 1 mg
Catalog # ACRBS2N-C52HD-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together